(±)-3-Amino-1,2-propanediol 98%
Supplier: Apollo Scientific
Danger
OR10134-100G OR10134-25G OR10134-500G
APOSOR10134-100GEA 50.9 EUR
APOSOR10134-100G APOSOR10134-25G APOSOR10134-500G
(±)-3-Amino-1,2-propanediol 98%
(±)-3-Amino-1,2-propanediol
Formula: C₃H₉NO₂
MW: 91.11 g/mol
Boiling Pt: 264…265 °C
Density: 1.175 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00008140
CAS Number: 616-30-8
EINECS: 210-475-8
UN: 2735
ADR: 8,II