1,3-Dibromo-5,5-dimethylhydantoin 98%
Supplier: Apollo Scientific
Danger
OR10135-2.5KG OR10135-500G OR10135-100G
APOSOR10135-2.5KGEA 241 EUR
APOSOR10135-2.5KG APOSOR10135-500G APOSOR10135-100G
1,3-Dibromo-5,5-dimethylhydantoin 98%
1,3-Dibromo-5,5-dimethylhydantoin
Formula: C₅H₆Br₂N₂O₂
Melting Pt: 197…199 °C (dec.)
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00003189
CAS Number: 77-48-5
EINECS: 201-030-9
UN: 3087
ADR: 5.1+6.1,II