2-Chlorothiophene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Thienyl chloride
OR10138-100G OR10138-5G OR10138-25G
APOSOR10138-100GEA 75.4 EUR
APOSOR10138-100G APOSOR10138-5G APOSOR10138-25G
2-Chlorothiophene 98%
2-Chlorothiophene
Formula: C₄H₃ClS
MW: 118.59 g/mol
Boiling Pt: 127…129 °C
Melting Pt: –72 °C
Density: 1.286 g/cm³ (25 °C)
Flash Pt: 23 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005421
CAS Number: 96-43-5
EINECS: 202-505-3
UN: 1992
ADR: 3+6.1 III