3-Thiophenemethanol 98+%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Thienylmethanol, Thiophen-3-ylmethanol, Thiophen-3-methanol
OR10141-1G OR10141-25G OR10141-5G
APOSOR10141-1GEA 21.9 EUR
APOSOR10141-1G APOSOR10141-25G APOSOR10141-5G
3-Thiophenemethanol 98+%
3-Thiophenemethanol
Formula: C₅H₆OS
MW: 114.17 g/mol
Boiling Pt: 86…88 °C/10mmHg
Density: 1.211 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00014534
CAS Number: 71637-34-8
EINECS: 275-741-8