2,3,5-Tribromothiophene
Supplier: Apollo Scientific
Warning
OR10149-100G OR10149-25G OR10149-500G
APOSOR10149-100GEA 46.4 EUR
APOSOR10149-100G APOSOR10149-25G APOSOR10149-500G
2,3,5-Tribromothiophene
2,3,5-Tribromothiophene
Formula: C₄HBr₃S
Boiling Pt: 120 °C/11mmHg
Melting Pt: 25…29 °C
Density: 2.483 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00014521
CAS Number: 3141-24-0
EINECS: 221-544-7