2-Bromo-5-nitrothiophene
Supplier: Apollo Scientific
Warning
OR10150-25G
APOSOR10150-25GEA 109 EUR
APOSOR10150-25G
2-Bromo-5-nitrothiophene
2-Bromo-5-nitrothiophene
Formula: C₄H₂BrNO₂S
MW: 208.04 g/mol
Boiling Pt: 235…237 °C
Melting Pt: 44…48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00022493
CAS Number: 13195-50-1
EINECS: 236-155-8