2-Chloro-3-methylthiophene
Supplier: Apollo Scientific
Danger
OR10152-100G OR10152-25G OR10152-500G
APOSOR10152-100GEA 58 EUR
APOSOR10152-100G APOSOR10152-25G APOSOR10152-500G
2-Chloro-3-methylthiophene
2-Chloro-3-methylthiophene
Formula: C₅H₅ClS
Boiling Pt: 147…149 °C
Density: 1.211 g/cm³ (25 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00130083
CAS Number: 14345-97-2
UN: 1993
ADR: 3,III