3-Methylthiophene-2-carboxaldehyde 90%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Methyl-2-thiophenecarboxaldehyde, 3-Methylthiophene-2-carbaldehyde
OR10155-100G OR10155-25G
APOSOR10155-100GEA 98.6 EUR
APOSOR10155-100G APOSOR10155-25G
3-Methylthiophene-2-carboxaldehyde 90%
3-Methylthiophene-2-carboxaldehyde
Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 215 °C
Density: 1.17 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005430
CAS Number: 5834-16-2
EINECS: 227-418-8