4-Bromo-2,6-dichlorobenzenesulphonamide
Supplier: Apollo Scientific
Warning
OR1016-10G OR1016-5G
APOSOR1016-10GEA 498 EUR
APOSOR1016-10G APOSOR1016-5G
4-Bromo-2,6-dichlorobenzenesulphonamide
4-Bromo-2,6-dichlorobenzenesulphonamide
Formula: C₆H₄BrCl₂NO₂S
Storage Temperature: Ambient
MDL Number: MFCD03094616
CAS Number: 351003-55-9