2,3,3-Trimethylindolenine
Supplier: Apollo Scientific
Warning
OR10172-100G OR10172-25G OR10172-500G
APOSOR10172-100GEA 59.6 EUR
APOSOR10172-100G APOSOR10172-25G APOSOR10172-500G
2,3,3-Trimethylindolenine
2,3,3-Trimethylindolenine
Formula: C₁₁H₁₃N
MW: 159.23 g/mol
Boiling Pt: 228…229 °C/744mmHg
Density: 0.992 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00005724
CAS Number: 1640-39-7