2-Methyl-1H-indole-3-carbaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 2-Methylindole-3-carboxaldehyde, 3-Formyl-2-methylindole
OR10175-1G OR10175-5G
APOSOR10175-1GEA 21.9 EUR
APOSOR10175-1G APOSOR10175-5G
2-Methyl-1H-indole-3-carbaldehyde
2-Methyl-1H-indole-3-carbaldehyde
Formula: C₁₀H₉NO
MW: 159.1846 g/mol
Melting Pt: 200…205 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012077
CAS Number: 5416-80-8
EINECS: 226-512-6