Indol-3-ylacetonitrile 98%
Supplier: Apollo Scientific
Warning
Synonyms: Indole-3-Acetonitrile, 3-Indolylacetonitrile, 2-(1H-Indol-3-yl)acetonitrile, 3-Indoleacetonitrile
OR10177-100G OR10177-25G OR10177-5G
APOSOR10177-100GEA 91.5 EUR
APOSOR10177-100G APOSOR10177-25G APOSOR10177-5G
Indol-3-ylacetonitrile 98%
Indol-3-ylacetonitrile
Formula: C₁₀H₈N₂
MW: 156.19 g/mol
Boiling Pt: 157…160 °C
Melting Pt: 33…36 °C
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005628
CAS Number: 771-51-7
EINECS: 212-232-1
UN: 3439
ADR: 6.1,III