Indole-5-carboxylic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1H-Indole-5-carboxylic acid
OR10183-25G OR10183-100G
APOSOR10183-25GEA 95.7 EUR
APOSOR10183-25G APOSOR10183-100G
Indole-5-carboxylic acid 98%
Indole-5-carboxylic acid
Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 208…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00005678
CAS Number: 1670-81-1
EINECS: 216-799-6