5-Methoxy-1H-indole-3-carbaldehyde
Supplier: Apollo Scientific
Warning
OR10185-25G OR10185-5G
APOSOR10185-25GEA 109 EUR
APOSOR10185-25G APOSOR10185-5G
5-Methoxy-1H-indole-3-carbaldehyde
5-Methoxy-1H-indole-3-carbaldehyde
Formula: C10H9NO2
MW: 175.19 g/mol
Melting Pt: 179…184 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005623
CAS Number: 10601-19-1
EINECS: 234-220-5