Indole
Supplier: Apollo Scientific
Danger
OR10189-100G OR10189-25G OR10189-500G
APOSOR10189-100GEA 46.5 EUR
APOSOR10189-100G APOSOR10189-25G APOSOR10189-500G
Indole
Indole
Formula: C₈H₇N
Boiling Pt: 253…254 °C
Melting Pt: 52…54 °C
Density: 1.22 g/cm³ (25 °C)
Flash Pt: 121 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005607
CAS Number: 120-72-9
EINECS: 204-420-7
UN: 2811
ADR: 6.1,III