Indole-3-acetamide
Supplier: Apollo Scientific
Warning
OR10210-100G OR10210-25G OR10210-5G
APOSOR10210-100GEA 95.7 EUR
APOSOR10210-100G APOSOR10210-25G APOSOR10210-5G
Indole-3-acetamide
Formula: C₁₀H₁₀N₂O
Melting Pt: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00005641
CAS Number: 879-37-8
EINECS: 212-904-4