2-Aminothiazole-5-carbaldehyde 96%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Aminothiazole-5-carboxaldehyde, 2-Amino-1,3-thiazole-5-carboxaldehyde
OR10318-25G OR10318-5G
APOSOR10318-25GEA 452 EUR
APOSOR10318-25G APOSOR10318-5G
2-Aminothiazole-5-carbaldehyde 96%
2-Aminothiazole-5-carbaldehyde
Formula: C₄H₄N₂OS
MW: 128.15 g/mol
Melting Pt: 160…166 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD02179570
CAS Number: 1003-61-8