6-Bromo-1-hexanol
Supplier: Apollo Scientific
Warning
Synonyms: 6-Bromohexan-1-ol, Hexamethylene bromohydrin, 1-Bromo-6-hydroxyhexane
OR10365-5G OR10365-25G OR10365-100G
APOSOR10365-5GEA 21.9 EUR
APOSOR10365-5G APOSOR10365-25G APOSOR10365-100G
6-Bromo-1-hexanol
6-Bromo-1-hexanol
Formula: BrH₂C(CH₂)₄CH₂OH
MW: 181.07 g/mol
Boiling Pt: 105…106 °C/5mmHg
Density: 1.384 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002983
CAS Number: 4286-55-9