3-(Bromomethyl)phenylboronic acid
Supplier: Apollo Scientific
Danger
Synonyms: 3-Bromomethylphenylboronic acid, 3-(Bromomethyl)benzeneboronic acid, Dihydroxy(3-(bromomethyl)phenyl)borane, (3-(Bromomethyl)phenyl)metaboric acid
OR10378-25G
APOSOR10378-25GEA 151 EUR
APOSOR10378-25G
3-(Bromomethyl)phenylboronic acid
3-(Bromomethyl)phenylboronic acid
Formula: C₇H₈BBrO₂
MW: 214.85 g/mol
Melting Pt: 216…228 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01632207
CAS Number: 51323-43-4
UN: 3261
ADR: 8,III