2-Chloro-1-(ethoxymethyl)imidazole
Supplier: Apollo Scientific
Warning
OR10410-100MG OR10410-1G OR10410-5G
APOSOR10410-100MGEA 50.9 EUR
APOSOR10410-100MG APOSOR10410-1G APOSOR10410-5G
2-Chloro-1-(ethoxymethyl)imidazole
2-Chloro-1-(ethoxymethyl)imidazole
Formula: C₆H₉ClN₂O
MW: 160.60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788569
CAS Number: 850429-55-9