3,5-Dibromotoluene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1,3-Dibromo-5-methylbenzene
OR10440-25G OR10440-100G
APOSOR10440-25GEA 21.9 EUR
APOSOR10440-25G APOSOR10440-100G
3,5-Dibromotoluene 97%
3,5-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 246 °C
Melting Pt: 37…41 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00013528
CAS Number: 1611-92-3