1-Methyl-1H-imidazole-4,5-dicarboxylic acid
Supplier: Apollo Scientific
Warning
Synonyms: 4,5-Dicarboxy-1-methyl-1H-imidazole, 1-Methylimidazole-4,5-dicarboxylic Acid
OR10441-1G OR10441-5G
APOSOR10441-1GEA 79.9 EUR
APOSOR10441-1G APOSOR10441-5G
1-Methyl-1H-imidazole-4,5-dicarboxylic acid
1-Methyl-1H-imidazole-4,5-dicarboxylic acid
Formula: C₆H₆N₂O₄
MW: 170.12 g/mol
Melting Pt: 262 °C
Storage Temperature: Ambient
MDL Number: MFCD01552045
CAS Number: 19485-38-2