3,5-Dichlorophenylboronic acid
Supplier: Apollo Scientific
Warning
OR10442-100G OR10442-25G OR10442-5G
APOSOR10442-100GEA 143 EUR
APOSOR10442-100G APOSOR10442-25G APOSOR10442-5G
3,5-Dichlorophenylboronic acid
3,5-Dichlorophenylboronic acid
Formula: C₆H₅BCl₂O₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00051935
CAS Number: 67492-50-6