2,4-Dichlorophenylboronic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: Dihydroxy(2,4-dichlorophenyl)borane, 2,4-Dichlorobenzeneboronic acid, (2,4-Dichlorophenyl)metaboric acid
OR10443-25G OR10443-5G OR10443-100G
APOSOR10443-25GEA 43.5 EUR
APOSOR10443-25G APOSOR10443-5G APOSOR10443-100G
2,4-Dichlorophenylboronic acid 98%
2,4-Dichlorophenylboronic acid
Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
Melting Pt: 246…249 °C
Storage Temperature: Ambient
MDL Number: MFCD00013930
CAS Number: 68716-47-2