Ethyl thiazole-4-carboxylate 98%
Supplier: Apollo Scientific
Warning
OR10476-25G
APOSOR10476-25GEA 97.3 EUR
APOSOR10476-25G
Ethyl thiazole-4-carboxylate 98%
Ethyl thiazole-4-carboxylate
Formula: C₆H₇NO₂S
MW: 157.1900 g/mol
Boiling Pt: 130 °C/12mmHg
Melting Pt: 51…55 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03788566
CAS Number: 14527-43-6