6-Hydroxy-2-naphthoic acid 97%

Warning

OR10498-100G OR10498-1KG OR10498-500G
APOSOR10498-100GEA 101 EUR
APOSOR10498-100G APOSOR10498-1KG APOSOR10498-500G
6-Hydroxy-2-naphthoic acid 97%
6-Hydroxy-2-naphthoic acid
Formula: C₁₁H₈O₃
MW: 188.18 g/mol
Melting Pt: 240…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00060070
CAS Number: 16712-64-4
EINECS: 240-759-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR