4-(Hydroxymethyl)phenylboronic acid 97+%
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Hydroxymethyl)benzeneboronic acid
OR10501-100G OR10501-25G OR10501-5G
APOSOR10501-100GEA 157 EUR
APOSOR10501-100G APOSOR10501-25G APOSOR10501-5G
4-(Hydroxymethyl)phenylboronic acid 97+%
4-(Hydroxymethyl)phenylboronic acid
Formula: C₇H₉BO₃
MW: 151.96 g/mol
Melting Pt: 251…256 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00792672
CAS Number: 59016-93-2