4-Methoxy-2-methylphenylboronic acid
Supplier: Apollo Scientific
Warning
Synonyms: 4-Methoxy-2-methylbenzeneboronic acid, (4-Methoxy-2-methylphenyl)boronic acid
OR10527-25G
APOSOR10527-25GEA 151 EUR
APOSOR10527-25G
4-Methoxy-2-methylphenylboronic acid
4-Methoxy-2-methylphenylboronic acid
Formula: C₈H₁₁BO₃
MW: 165.99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02684315
CAS Number: 208399-66-0