Methyl indole-6-carboxylate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 1H-indole-6-carboxylate
OR10544-5G OR10544-25G OR10544-100G
APOSOR10544-5GEA 21.9 EUR
APOSOR10544-5G APOSOR10544-25G APOSOR10544-100G
Methyl indole-6-carboxylate 98%
Methyl indole-6-carboxylate
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 76…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00211063
CAS Number: 50820-65-0