Methyl tetrahydropyran-4-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl tetrahydro-2H-pyran-4-carboxylate
OR10552-100G OR10552-25G OR10552-500G
APOSOR10552-100GEA 144 EUR
APOSOR10552-100G APOSOR10552-25G APOSOR10552-500G
Methyl tetrahydropyran-4-carboxylate
Methyl tetrahydropyran-4-carboxylate
Formula: C₇H₁₂O₃
MW: 144.1684 g/mol
Boiling Pt: 196.8 °C
Melting Pt: –33 °C
Density: 1.08 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD01075170
CAS Number: 110238-91-0