Thiazole-5-carboxyaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 5-Thiazolecarboxaldehyde, 5-Formylthiazole
OR10588-1G OR10588-5G
APOSOR10588-1GEA 30.6 EUR
APOSOR10588-1G APOSOR10588-5G
Thiazole-5-carboxyaldehyde
Thiazole-5-carboxyaldehyde
Formula: C₄H₃NOS
Boiling Pt: 92…94 °C/16mmHg
Density: 1.304 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD02179516
CAS Number: 1003-32-3