2-Amino-6-chloropurine 99%
Supplier: Apollo Scientific
Warning
Synonyms: 6-Chloroguanine, 6-Chloro-7H-purin-2-ylamine
OR10600-100G OR10600-25G OR10600-500G
APOSOR10600-100GEA 31.9 EUR
APOSOR10600-100G APOSOR10600-25G APOSOR10600-500G
2-Amino-6-chloropurine 99%
2-Amino-6-chloropurine
Formula: C₅H₄ClN₅
MW: 169.57 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00075252
CAS Number: 10310-21-1
EINECS: 233-686-7