(4-Carboxybutyl)triphenylphosphonium bromide 98%
Supplier: Apollo Scientific
Warning
APOSOR10636-25GEA 59.7 EUR
APOSOR10636-25G APOSOR10636-500G APOSOR10636-100G
(4-Carboxybutyl)triphenylphosphonium bromide 98%
(4-Carboxybutyl)triphenylphosphonium bromide
Formula: C₂₃H₂₄BrO₂P
MW: 443,32 g/mol
Melting Pt: 204…209 °C
Flash Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00011906
EINECS: 241-782-5