4-tert-Butylthiophenol
Supplier: Apollo Scientific
Warning
OR10686-25G OR10686-5G OR10686-1G
APOSOR10686-25GEA 94.4 EUR
APOSOR10686-25G APOSOR10686-5G APOSOR10686-1G
4-tert-Butylthiophenol
4-tert-Butylthiophenol
Formula: C₁₀H₁₄S
Boiling Pt: 238 °C
Melting Pt: –11 °C
Density: 0.99 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00022067
CAS Number: 2396-68-1
EINECS: 219-255-6