2-Bromo-p-toluidine
Supplier: Apollo Scientific
Danger
Synonyms: 2-Bromo-4-methylaniline, 4-Amino-3-bromotoluene
OR1073-100G OR1073-25G
APOSOR1073-100GEA 43.5 EUR
APOSOR1073-100G APOSOR1073-25G
2-Bromo-p-toluidine
2-Bromo-p-toluidine
Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 120…122 °C/30mmHg
Melting Pt: 14…16 °C
Density: 1.5 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007635
CAS Number: 583-68-6
EINECS: 209-515-7
UN: 2810
ADR: 6.1 III