4-Bromo-2-methylanisole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-2-methoxytoluene, 1-Bromo-4-methoxy-3-methylbenzene, 4-Bromo-1-methoxy-2-methylbenzene
OR1076-100G
APOSOR1076-100GEA 98.6 EUR
APOSOR1076-100G
4-Bromo-2-methylanisole 98%
4-Bromo-2-methylanisole
Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 124…125 °C/25mmHg
Melting Pt: 66…69 °C
Storage Temperature: Ambient
MDL Number: MFCD01321139
CAS Number: 14804-31-0