4-Chloro-2-iodoaniline
Supplier: Apollo Scientific
Danger
OR1082-100G OR1082-25G OR1082-10G
APOSOR1082-100GEA 81.2 EUR
APOSOR1082-100G APOSOR1082-25G APOSOR1082-10G
4-Chloro-2-iodoaniline
4-Chloro-2-iodoaniline
Formula: C₆H₅ClIN
Melting Pt: 39…43 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863737
CAS Number: 63069-48-7
EINECS: 263-839-3
UN: 2811
ADR: 6.1,III