5-Bromobenzofuran
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromobenzo[b]furan
OR10831-10G OR10831-1G OR10831-5G
APOSOR10831-10GEA 53.8 EUR
APOSOR10831-10G APOSOR10831-1G APOSOR10831-5G
5-Bromobenzofuran
5-Bromobenzofuran
Formula: C₈H₅BrO
MW: 197.04 g/mol
Boiling Pt: 260 °C
Melting Pt: 8 °C
Density: 1.593 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03407317
CAS Number: 23145-07-5