2-Chloroethyl phenyl sulphide 98%
Supplier: Apollo Scientific
Danger
OR10842-25G
APOSOR10842-25GEA 121 EUR
APOSOR10842-25G
2-Chloroethyl phenyl sulphide 98%
2-Chloroethyl phenyl sulphide
Formula: C₈H₉ClS
MW: 172.68 g/mol
Boiling Pt: 245 °C
Density: 1.178 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00013691
CAS Number: 5535-49-9
EINECS: 226-891-8
UN: 2922
ADR: 8+6.1,III