Iodobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: Phenyliodide
OR1085-100G OR1085-500G OR1085-25G
APOSOR1085-100GEA 50.9 EUR
APOSOR1085-100G APOSOR1085-500G APOSOR1085-25G
Iodobenzene 98%
Iodobenzene
Formula: C₆H₅I
Boiling Pt: 188…189 °C
Melting Pt: –31 °C
Density: 1.826 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001029
CAS Number: 591-50-4
EINECS: 209-719-6