2,5-Dichlorobenzaldehyde 98%
Supplier: Apollo Scientific
Warning
OR10904-5G OR10904-25G OR10904-100G
APOSOR10904-5GEA 21.9 EUR
APOSOR10904-5G APOSOR10904-25G APOSOR10904-100G
2,5-Dichlorobenzaldehyde 98%
2,5-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 231…233 °C
Melting Pt: 53…58 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00156140
CAS Number: 6361-23-5