3-Chlorobenzoic acid
Supplier: Apollo Scientific
Warning
OR10911-100G OR10911-25G OR10911-500G
APOSOR10911-100GEA 29 EUR
APOSOR10911-100G APOSOR10911-25G APOSOR10911-500G
3-Chlorobenzoic acid
3-Chlorobenzoic acid
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 281
Melting Pt: 153…157 °C
Density: 1.496 g/cm³ (25 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00002491
CAS Number: 535-80-8
EINECS: 208-618-4
Merck Index: 13,02142