15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) 98%
Supplier: Apollo Scientific
Warning
Synonyms: 15-Crown-5, 1,4,7,10,13-Pentaoxacyclopentadecane
OR10918-100G OR10918-25G OR10918-5G
APOSOR10918-100GEA 109 EUR
APOSOR10918-100G APOSOR10918-25G APOSOR10918-5G
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) 98%
15-Crown-5 (1,4,7,10,13-Pentaoxacyclopentadecane, Lead ionophore V)
Formula: C₁₀H₂₀O₅
MW: 220.27 g/mol
Boiling Pt: 150…155 °C
Density: 1.109 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005110
CAS Number: 33100-27-5
EINECS: 251-379-6