Dibenzo-30-crown-10 (dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriacontane)
Supplier: Apollo Scientific
Warning
OR10923-1G OR10923-5G
APOSOR10923-1GEA 202 EUR
APOSOR10923-1G APOSOR10923-5G
Dibenzo-30-crown-10 (dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriacontane)
Dibenzo-30-crown-10 (Dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriacontane)
Formula: C₂₈H₄₀O₁₀
Melting Pt: 104…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00054624
CAS Number: 17455-25-3