4-Chloro-6-methoxypyrimidin-2-amine 96%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-4-chloro-6-methoxypyrimidine
OR10959-100G OR10959-25G
APOSOR10959-100GEA 29 EUR
APOSOR10959-100G APOSOR10959-25G
4-Chloro-6-methoxypyrimidin-2-amine 96%
4-Chloro-6-methoxypyrimidin-2-amine
Formula: C₅H₆ClN₃O
MW: 159.57 g/mol
Melting Pt: 168…176 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00233535
CAS Number: 5734-64-5
EINECS: 410-050-9