2-Phenylaniline 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Aminobiphenyl, Biphenyl-2-ylamine, 2-Biphenylylamine
OR10963-25G OR10963-100G OR10963-5G
APOSOR10963-25GEA 137 EUR
APOSOR10963-25G APOSOR10963-100G APOSOR10963-5G
2-Phenylaniline 98%
2-Phenylaniline
Formula: C₁₂H₁₁N
MW: 169.23 g/mol
Boiling Pt: 299 °C
Melting Pt: 47…50 °C
Flash Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD00007701
CAS Number: 90-41-5