N,N'-Bis(2-aminoethyl)-1,3-propanediamine
Supplier: Apollo Scientific
Danger
Synonyms: 3,7-Diaza-1,9-nonanediamine, N1,N1'-(Propane-1,3-diyl)bis(ethane-1,2-diamine), 1,4,8,11-Tetraazaundecane
OR10977-25G OR10977-5G OR10977-100G
APOSOR10977-25GEA 60.9 EUR
APOSOR10977-25G APOSOR10977-5G APOSOR10977-100G
N,N'-Bis(2-aminoethyl)-1,3-propanediamine
N,N'-Bis(2-aminoethyl)-1,3-propanediamine
Formula: C₇H₂₀N₄
MW: 160.27 g/mol
Boiling Pt: 280 °C
Density: 0.96 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00008174
CAS Number: 4741-99-5
UN: 2735
ADR: 8,III