Ethyl-4-iodobenzoate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 4-iodobenzoate, 4-Iodobenzoic acid ethyl ester
OR10986-25G OR10986-100G OR10986-500G
APOSOR10986-25GEA 23.2 EUR
APOSOR10986-25G APOSOR10986-100G APOSOR10986-500G
Ethyl-4-iodobenzoate 98%
Ethyl 4-iodobenzoate
Formula: C₉H₉IO₂
MW: 276.07 g/mol
Boiling Pt: 281…283 °C
Density: 1.65 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00017344
CAS Number: 51934-41-9