4-Iodo-1H-pyrazole 99%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Iodopyrazole
OR10990-100G OR10990-25G OR10990-500G
APOSOR10990-100GEA 39.3 EUR
APOSOR10990-100G APOSOR10990-25G APOSOR10990-500G
4-Iodo-1H-pyrazole 99%
4-Iodo-1H-pyrazole
Formula: C₃H₃IN₂
MW: 193.97 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005244
CAS Number: 3469-69-0
EINECS: 222-434-1