5-(Chloromethyl)-3-(4-methoxyphenyl)-1,2,4-oxadiazole
Supplier: Apollo Scientific
OR110030-1G OR110030-250MG
APOSOR110030-1GEA 0 EUR
APOSOR110030-1G APOSOR110030-250MG
5-(Chloromethyl)-3-(4-methoxyphenyl)-1,2,4-oxadiazole
5-(Chloromethyl)-3-(4-methoxyphenyl)-1,2,4-oxadiazole
Formula: C₁₀H₉ClN₂O₂
Melting Pt: 42…44 °C
Storage Temperature: Ambient
CAS Number: 57238-76-3